Orders LinkContactUsLinkComponent
HighPressureFlexibleTubingHeader.jpg
High Pressure Flexible Tubing
If your research or biopharma process involves bulk transfers, you’re likely familiar with the complications that come with typical high-pressure tubing. Most tubing designed for high-pressure is not designed to meet regulations for Pharmaceutical or Food & Beverage industries, or the low-extractable, chemical resistance, precision, and high-performance needed.
High Pressure Flexible Tubing | Avantor
Masterflex High Pressure Flexible Tubing is the Answer
Does your research require you to use peristaltic pumps at pressures of up to 100 psi? Then our Masterflex® High-Pressure flexible plastic tubing is what you need.
Masterflex® L/S® high-pressure precision process pump system, which transfers fluids at continuous pressures up to 125 psi (8.5 bar), featuring a flow range of 0.9 to 90 ml/min (depends on drive rpm and tubing size)
Choose from two formulations and three sizes:
MasterflexTygon295.jpg
Masterflex® Tygon® A-60-F (Norprene) High Pressure Pump Tubing
  • L/S® Tygon® A-60-F, 16HP size tubing (MFLX06504-16), capable of delivering 100 psi maximum continuous pressure and 125 psi intermittent pressure, with a flow range of 0.9 to 90 mL/min.
  • L/S® Tygon® A-60-F, 15HP size tubing (MFLX06504-15), capable of a flow range of 1.7 to 170 mL/min, with 80 psi maximum continuous pressure and 100 psi maximum intermittent pressure. This flexible high-pressure tubing is easily adaptable to a wide variety of research applications.
Learn more about Norprene (A 60 G) tubing characteristics
MasterflexPharMed295.jpg
Masterflex® PharMed® BPT High Pressure Pump Tubing
Nontoxic, nonhemolytic, extremely low permeability
  • L/S® PharMed® BPT, 16HP size tubing (MFLX05664-16), Max. flow rate 90 ml/min, Max.pressure 8.5 bar (125 psi)
  • L/S® PharMed® BPT, 24HP size tubing (MFLX05664-24), Max. flow rate 240 ml/min, Max. pressure 5.5 bar (80 psi)
Learn more about PharMed tubing characteristics
Typical Applications of Masterflex High Pressure Flexible Tubing
Masterflex flexible tubing is good for a variety of high-pressure applications such as:
  • Cosmetics production
  • Cell and tissue culture work
  • Pharmaceutical research
  • Biotechnology and biologics
  • Water treatment
  • Food and beverage development
  • Environmental preservation
Choosing the Right Masterflex High Pressure Flexible Tubing
Make sure you get the high-pressure flexible tubing you need to keep your lab running like clockwork.
Contact us for more information today