Orders LinkContactUsLinkComponent
 Guide to Liquid Pumps | Avantor
Guide to Liquid Pumps
Use the guide below to help you select the best type of pump for your application. This information is intended as a general guideline and will not hold true for all pumps within a classification; check individual pump specifications on the given product pages for complete details.
Pump type
Max flow
ranges
Max
pressure
Self-
priming
Pulseless
flow
Fluid
viscosity
Particulate
matter
Run
dry
Advantages
GPM
L/min
Air-operated
diaphragm
5.0 to
225
19.0 to
851
Up to
125 psi
Excellent
Fair
Medium
Yes
Yes
Use for highly viscous or particulate-laden
fluids and where electricity is not available
Bellows
0.008 to
26.4
0.03 to
100
Up to
73 psi
Good
Poor
Medium
Yes
Yes
Can pump liquids or gases
Centrifugal
2.3 to
1200
8.7 to
4542
Up to
275 psi
Poor
Excellent
Light
No
No
Fluid transfer at high flow rates
and low pressures
Diaphragm
0.003 to
5.2
0.01 to
19.7
Up to
300 psi
Good
Poor
Medium
No
Yes
High-accuracy; for applications
such as pH/ORP control
Double-
diaphragm
1.0 to
4.0
3.79 to
15.1
Up to
95 psi
Excellent
Fair
Medium
Yes
Yes
Use for viscous or
particulate-laden fluids
Drum
6.0 to
63.0
22.7 to
238
Up to
35 psi
Poor
Excellent
Medium
No
No
Economical. Used to pump fluids from drums and cannisters
Flexible
impeller
3.8 to
50.0
14.4 to
189
Up to
60 psi
Excellent
Excellent
Light
No
No
Low-cost utility pump
Flexible
liner
1.0 to
10.0
3.8 to
37.8
Up to
50 psi
Excellent
Excellent
Medium
Yes
Yes
Gentle pumping action uses no
seals—pulseless, can run dry
Gear
0.006 to
74.0
0.026 to
280
Up to
1500 psi
Poor
Excellent
Medium
No
No
Pulseless flow at high pressures
Hand
2.0 to
48
7.6 to
181.7
Good
Fair
Light
No
No
Economical. No electrical parts
Hose
0.66 to
68
2.5
257.4
Up to
220 psi
Excellent
Poor
Medium
Yes
Yes
Noncontaminating; Use for high flow and
high pressure applications
Piston
0.004 to
107
0.015 to
405
Up to
5000 psi
Good
Poor
Medium
No
Yes
Highest pressure and accuracy;
ideal for HPLC applications
Pressure
Washer
2.0 to
4.0
7.6 to
15.1
Up to
3000 psi
N/A
Excellent
Light
No
No
High pressure capabilities;
portability, heating
Progressing
cavity
0.5 to
13
1.9 to
49
Up to
100 psi
Fair
Excellent
Very
heavy
Yes
No
Pulseless flow for highly viscous
or particulate-laden fluids
Rotary vane
0.75 to
4.3
2.8 to
16.3
Up to
240 psi
Fair
Very
good
Light
No
No
High-pressure capabilities;
low shear
Submersible
3.0 to
180
11.4 to
681.3
Up to
50 psi
Poor
Excellent
Light
No
No
Use for emptying tanks or sumps
Syringe
0.002 to
0.04
0.008 to
0.15
Up to
40 psi
N/A
Excellent
Light
No
Yes
Low flow rates at high pressure and accuracy