Orders LinkContactUsLinkComponent
 Ismatec: Tubing Selection Guide | Avantor
Ismatec: Tubing Selection Guide
Selecting Your Tubing
To complete your Ismatec pumping system, order two- or three-stop color-coded tubing or nominal tubing. A variety of materials and sizes are available, as well as extension tubing.
Tubing Material Information
MaterialApperanceTemperatureLife at 100 rpmAdvantagesApplication
TygonClear-50 to 74°C300 hoursClear and economicalAcids, alkalies, viscous fluids
Tygon LFLClear-50 to 74°C300 hoursPharmaceutical grade.Longest flex life of all TygonAcids, alkalies, viscous fluids, pharmaceuticals
Silicone (peroxide cured)Translucent-60 to 260°C800 hoursExcellent biocompatibilitySterile fluids
Silicone (platinum cured)Translucent-60 to 260°C800 hoursVery low leaching of organic compoundsSterile fluids
NorpreneOpaque, black-50 to 130°C1400 hoursLong lifeAcids, alkalies, general purpose
VitonOpaque, black-20 to 205°C160 hoursResists most chemicalsAcids, alkalies, solvents, viscous fluids
PharMedOpaque, beige-50 to 130°C1400 hoursPharmaceutical gradeAcids, alkalies, sterile fluids, viscous fluids
How to order 2- and 3- stop tubing
Choose either 2- or 3- stop tubing per your pump’s requirements
Tubing Types
TubingTypesImg1.jpg
Two-Stop Tubing
Use two-stop tubing in standard-sized cartridges and tubing bed sets. Choose from the seven tubing material formulations below. Extension tubing is also available. Tubing stops are color-coded according to tubing ID for quick identification. Stops hold the tubing at the optimal tension necessary for pump head rollers to occlude the tubing. The result is reliable pumping and repeatable flow rates.
TubingTypesImg2.jpg
Three-Stop Tubing
Use three-stop tubing in minicartridges. Choose from the seven tubing material formulations below. Extension tubing is also available. Tubing stops, color-coded according to tubing ID, hold the tubing at the optimal tension necessary for pump head rollers to occlude the tubing. The result is reliable pumping and repeatable flow rates. Three stops give you two sections of tubing to use instead of just one. When one section of tubing fatigues, a fresh section is ready to use.
TubingTypesImg3.jpg
Nominal Tubing
Use nominal tubing with the quick-change and tubing bed pumps. Choose from seven tubing material formulations below.
Step1.jpg
STEP 1
Choose a tubing material
Step2.jpg
STEP 2
Choose the ID size needed per flow rate and pump system
STEP 3
Complete your system with extension tubing. You can connect stopped tubing and extension tubing with hose barb fittings or by splicing together.