Orders LinkContactUsLinkComponent
Fluid Fundamentals | Avantor
Fluid Fundamentals
Fluids include liquids, gases, and mixtures of liquids, solids, and gases. For the purposes of this reference book, the terms fluid and liquid are both used to mean a pure liquid or a liquid mixed with gases or solids that acts essentially like a liquid in a pumping application.
Density or specific weight of a fluid is its weight per unit volume, often expressed in units of grams per cubic centimeter or pounds per cubic foot. The density of a fluid changes with temperature.
Example:
If weight is 36.36 kg (80 lb), density is 1.28 g/cm3 (80 lb/cu ft).
cube1.jpg
Specific gravity of a fluid is the ratio of its density to the density of water. Since it is a ratio, it has no units associated with it.
Example:
Specific gravity = 80 lb/62.4 lb or, Specific gravity = 1.282
Cube2.jpg