Orders LinkContactUsLinkComponent
Viral-Contamination1920-760.png
Viral Contamination in Biomanufacturing
Viral Contamination in Biomanufacturing | Avantor
What is viral contamination in biomanufacturing?
Viral contamination in biomanufacturing occurs when unwanted viruses enter the production process for vaccines, therapeutic proteins and other biopharmaceuticals. This can jeopardize the quality and safety of the final product and pose severe risks to humans who use those products.
Only small quantities of viral contaminants can significantly impact a product's efficacy and safety. Adopting strict monitoring and control measures is important to prevent and identify viral contamination. These measures help maintain the integrity of biomanufacturing processes and ensure the reliability of pharmaceutical products.
Identifying viral contamination
Detecting viral contamination is crucial to maintaining the quality of biomanufacturing processes. Viruses can enter production through many sources, such as contaminated raw materials or poor aseptic practices. Scientists can use several techniques to detect and identify viral contamination accurately so possible threats can be immediately addressed.
Raw material contamination
Raw material contamination is a critical concern in biopharmaceutical production. Detecting viral contaminants at an early stage is essential to avoid compromising product quality and ensure the production of safe and effective biopharmaceuticals.
Biomanufacturers can use several techniques, such as polymerase chain reaction (PCR) and enzyme-linked immunosorbent assay (ELISA). PCR is a molecular biology method that amplifies specific DNA sequences to help detect sensitive viral genetic material. ELISA uses antibodies to identify and quantify the presence of viruses by measuring their immune response. These techniques are robust tools for screening raw materials and can help manufacturers identify and eliminate viral contaminants before they impact the biomanufacturing process.
Equipment and facility contamination
Equipment sterility and cleanliness are vital in pharmaceutical and biotechnology manufacturing. Industry groups and government regulatory agencies often require stringent monitoring protocols to address this concern.
Swab testing is a commonly used method for regular assessments that require samples from critical surfaces and equipment. The samples are analyzed to detect viral particles and identify potential contaminants before they can compromise the manufacturing process or the final products.
Advanced imaging technology, such as high-resolution cameras and imaging systems, can augment traditional monitoring methods. This technology can help visually inspect equipment and facilities at microscopic levels. The combination of viral swab testing and advanced imaging technologies helps create a defense against contamination of equipment and facilities.
Human-induced contamination
Human-induced contamination, particularly involving biological materials, poses a significant risk of viral contamination. This occurs when people unintentionally introduce viral particles during the manufacturing process. It is important to monitor the health of scientists and other personnel closely. Regular health assessments can help identify individuals who could be carrying viruses.
It’s also important to implement strict hygiene protocols to prevent transmitting viruses in controlled environments. These protocols include using personal protective equipment (PPE), practicing proper hand hygiene and following established cleanliness standards. Advanced detection techniques, such as nucleic acid-based assays and viral culture methods, can also help identify human-induced contamination. Nucleic acid-based assays, such as PCR and other molecular biology techniques, help identify specific viral genetic material. Organizations can identify and gain insights into viral contamination by combining these sophisticated methods.
Airborne contamination
The presence of harmful particles in the air causes airborne contamination. Advanced air quality monitoring systems can help detect and analyze airborne contaminants. These systems employ tools such as air samplers and particle counters to gather and measure the concentration of contaminants in the air.
Air samplers draw air from specific locations to check for virus-carrying particles. Particle counters, on the other hand, measure the number and size distribution of air particles. Molecular biology techniques can help identify specific viral strains within airborne contaminants with reverse transcription polymerase chain reaction (RT-PCR). RT-PCR allows scientists to detect and amplify the virus’ genetic material to help identify viral strains.
Process intermediates and final product monitoring
In biopharma manufacturing, closely monitoring different stages of the production process is essential. This requires using advanced techniques in which virus-specific assays play a significant role. These assays are designed to identify and measure viral presence throughout manufacturing. Scientists use highly sensitive and selective methods like PCR or ELISA to detect and quantify viruses at different production stages. This careful monitoring helps ensure compliance with regulatory standards and is a proactive measure to prevent contamination.
Integrating next-generation sequencing (NGS) enhances monitoring capabilities in biopharma manufacturing. NGS allows comprehensive analysis of genetic material in samples, providing scientists with information into the entire genomic composition. This high-throughput sequencing technology can identify potential contaminants, mutations or variations in genetic material.
Scientific techniques for prevention
Using specific prevention techniques to avoid contamination in biopharmaceutical manufacturing and research is essential. Aseptic techniques can help maintain clean environments and reduce the chance of introducing microorganisms to production settings. Some of these practices include proper hand washing, using sterile equipment or working inside a laminar flow hood.
Preventing contamination is especially important in fields that require precision and replicability, such as microbiology, cell culture and molecular biology. Aseptic techniques are also essential in viral research, where a sterile environment is vital. Contamination from other samples or external viral particles can affect experimental accuracy and damage the reliability of research.
Aseptic techniques can also protect researchers and prevent the accidental release of potentially harmful viruses in virology. Strictly adhering to these laboratory practices helps safeguard the integrity of the research or manufacturing and the safety of the personnel involved.
Additionally, using specialized equipment such as biosafety cabinets and PPE further enhances preventive measures. Biosafety cabinets create a contained environment and prevent viral particles from escaping, while PPE is an additional barrier against contamination. Comprehensive training programs and awareness initiatives for laboratory personnel regarding the importance of aseptic techniques can help prevent viral contamination. Regular equipment maintenance and validation also contribute to a robust defense against contamination.
Analyzing the results
Assessing contamination risks in biomanufacturing requires statistical methods, particularly for estimating the probability of viral contamination. One technique is to analyze historical data and quantify how likely contamination events are to occur.
By looking at previous incidents, process parameters and environmental conditions, statistical models can detect patterns and risk factors related to contamination. Statistical models can provide quantitative risk measures that allow biomanufacturers to focus on effective mitigation efforts.
One useful statistical tool is Bayesian inference. This technique uses prior knowledge and beliefs about contamination risks in the analysis to allow a more refined estimate of the probability of viral contamination. This technique is beneficial when data is scarce or uncertain because biomanufacturers can make informed decisions based on available evidence.
Monte Carlo simulations offer comprehensive techniques for assessing contamination risks in biomanufacturing processes. These simulations create many iterations of a model using random inputs to estimate the probability of contamination under different scenarios. By considering the uncertainties from process variability and environmental conditions, Monte Carlo simulations can estimate contamination risks and how they affect product quality and reliability. Biopharma manufacturers can use quantitative methods to evaluate contamination hazards and apply quality control measures that lower the likelihood of contamination incidents.
By using statistical techniques such as past data analysis, Bayesian reasoning and Monte Carlo models, biomanufacturers can anticipate and control risks and help guarantee the safety and quality of biopharmaceutical and biological products.
​​​​Future directions in viral contamination research
Preventing viral contamination in biopharmaceutical production requires a comprehensive approach that addresses multiple challenges. Among them are developing new technologies for rapid virus detection, improving cleaning methods for better decontamination and implementing real-time monitoring systems to detect contamination early. Researchers are conducting thorough evaluations of viral risks and developing effective strategies to manage those risks. They are investigating vaccines and treatments to deal with potential contamination problems. They are working to integrate different approaches to help enhance the safety, reliability and sustainability of biomanufacturing. Ongoing research could help enhance the sensitivity and precision of virus detection methods, create more effective techniques for removing viruses and increase our knowledge of viruses' behavior in biomanufacturing processes. That is why cooperation among industry players, regulatory bodies and academic institutions is vital to stimulating innovation and ensuring biopharmaceutical products' continued safety and quality.
Watch this webinar for more detailed information about new approaches to purification and viral inactivation of monoclonal antibodies.