Orders LinkContactUsLinkComponent
Recombinant-Proteins-hero.jpg
An Introduction to Recombinant Proteins
Explore the science behind recombinant proteins — from what it is to the types of recombinant proteins — and learn how they are revolutionizing biotechnology and medicine.
An Introduction to Recombinant Proteins | Avantor
Recombinant proteins are a vital aspect of modern biotechnology and have revolutionized various fields, such as medicine, agriculture and industrial processing. These proteins are used extensively in developing therapeutic drugs, producing enzymes for industrial processes and creating genetically modified organisms for agricultural purposes. This article aims to provide a comprehensive introduction to recombinant proteins, their production and development, and their associated challenges.
What is a recombinant protein?
Recombinant proteins are artificially produced by combining genetic material from different sources. Recombinant refers to recombining or joining genetic material to create a new combination that does not occur naturally.
To create a recombinant protein, scientists use genetic engineering techniques to insert a specific gene or DNA sequence that encodes a desired protein into the genome of a host organism, such as bacteria, yeast or mammalian cells. The host organism then serves as a factory for producing the protein of interest using its cellular machinery.
The process involves several steps. First, the gene encoding the desired protein is isolated and inserted into a DNA molecule called a vector. The vector acts as a carrier that can deliver the gene into the host organism’s cells. The vector contains additional genetic elements, such as promoter sequences, that allow the researchers to control the expression of the inserted gene.
Once the vector carrying the gene is introduced into the host organism, it integrates into its genome or exists separately as an episome. The host organism’s cellular machinery reads the gene’s DNA sequence and synthesizes the corresponding mRNA molecule. The mRNA is then translated into the protein by ribosomes, which further assemble amino acids in the order specified by the gene.
Why are recombinant proteins important?
Recombinant proteins are essential, as they facilitate the production of therapeutic proteins, enhance supply and safety, allow for customization and engineering, drive research and drug discovery, and find applications in various industrial sectors.
In the biopharma industry, these proteins form the basis for developing advanced biologic drugs, including monoclonal antibodies and recombinant vaccines, providing precise and effective treatments for diverse diseases.
Production of therapeutic proteins
Recombinant proteins are a valuable source for producing therapeutic proteins used in medicine. Many important proteins, such as insulin, growth factors, hormones, antibodies and clotting factors, are usually synthesized in large quantities using recombinant DNA technology.
Recombinant proteins have revolutionized the treatment of various diseases. They enable the production of synthetic insulin for diabetes control and targeted therapies like monoclonal antibodies for precise cancer treatment. In genetic disorders, they offer potential treatments by delivering therapeutic proteins or correcting faulty genes.
Furthermore, in immune-related conditions, the therapeutical recombinant proteins provide relief by modulating the immune response. It benefits patients with autoimmune diseases and other immune disorders.
Increased supply and availability
Recombinant protein production facilitates a scalable and reliable source of proteins that are otherwise scarce or challenging to extract from natural sources. Traditional protein isolation methods, such as purification from animal tissues, often yield limited quantities. Recombinant DNA technology enables the production of large quantities of proteins in a controlled and reproducible manner, ensuring a consistent supply for research, diagnostics and therapeutic applications.
Enhanced safety and purity
Recombinant proteins are safe compared to proteins obtained from natural sources. When scientists use recombinant DNA technology, they can tightly control the production process to reduce the chance of impurities, pathogens or toxins contaminating the final product. It further reduces the potential adverse reactions or side effects in patients receiving therapeutic proteins.
Customization and engineering
Recombinant proteins offer various opportunities with advancements in protein customization and engineering. Researchers can introduce specific modifications or alterations to enhance protein stability, functionality, or pharmacokinetic properties by manipulating the DNA sequence encoding the protein. It enables the development of improved protein variants with enhanced therapeutic efficacy, reduced immunogenicity or prolonged half-life in the body.
Research and drug discovery
These artificially produced proteins are also important in research and drug discovery. They allow scientists to delve into protein structure, function and interactions to understand biological processes and disease mechanisms better.
These versatile proteins find applications in various experimental techniques, including protein purification, crystallization, structural biology, enzymology and drug screening assays. They help identify and design potential therapeutic compounds for drug development. Additionally, they facilitate disease detection and monitoring in diagnostic tests.
Furthermore, researchers can understand complex molecular mechanisms, explore biological pathways and develop therapeutic approaches. Their versatility and broad applications make them invaluable for advancing scientific knowledge, expediting drug discovery and enhancing diagnostics.
Industrial applications
Recombinant proteins find applications across multiple industrial processes. One significant application lies in producing industrial enzymes using recombinant DNA technology. These enzymes play a crucial role in manufacturing food, beverages, textiles, detergents and biofuels.
The enzymes exhibit enhanced efficiency, enabling more streamlined and productive industrial processes. This leads to higher quality and purity of end products as their specificity is improved to target the substrates. Moreover, the enzymes demonstrate superior stability, allowing for longer and more reliable usage in industrial settings.
Industries can achieve cost-effective and sustainable practices by harnessing the potential of recombinant proteins. The heightened performance of these enzymes contributes to increased productivity, reduced waste and improved resource utilization.
How are recombinant proteins produced?
Recombinant proteins are produced through a step-by-step procedure involving gene isolation, DNA cloning, transformation, selection, protein expression, purification, quality control and scale-up production. These processes allow for the efficient and controlled production of recombinant proteins. Here’s a detailed explanation of how recombinant proteins are produced:
Gene isolation
The first step is identifying and isolating the gene that encodes the desired protein. It is usually done by various methods, such as identifying the gene from a natural source or synthesizing it using known protein sequences.
DNA cloning
Once the gene is isolated, it is inserted into a small, circular DNA molecule called a vector. As discussed before, vectors are commonly derived from plasmids that naturally occur in bacteria and can be easily manipulated in the laboratory. The gene is inserted into the vector at a specific location using specialized enzymes called restriction enzymes.
Transformation
The recombinant DNA molecule, which now contains the gene of interest within the vector, is introduced into host cells. Host cells can be bacteria, yeast or mammalian cells, depending on the desired protein and its functional requirements. This process, called transformation, allows the host cells to take up and incorporate the recombinant DNA into their genetic material.
Selection and expression
After transformation, the host cells are subjected to selective pressure to identify those who have successfully taken up the recombinant DNA. This selection is done by incorporating a selectable marker gene (antibiotic resistance gene) into the vector. Only cells that have successfully incorporated the recombinant DNA will survive in the presence of the specific antibiotic.
Protein production
Once the transformed cells are selected, they undergo the protein expression process. The recombinant gene in the host cells is transcribed into messenger RNA (mRNA), which carries the instructions for protein synthesis. The mRNA is then translated by the host cell’s ribosomes, producing the desired recombinant protein.
Protein purification
In protein purification, the host cells are harvested to obtain pure recombinant protein, and the protein of interest is purified from the cellular mixture. The purification process typically involves multiple steps, including chromatography techniques such as affinity, ion exchange and size exclusion chromatography to separate the recombinant protein from other cellular components, and filtration or dialysis for further purification and concentration.
Quality control
Once the recombinant protein is purified, it undergoes rigorous quality control tests to ensure its identity, purity and functionality. Depending on the specific protein and its intended use, these tests may include protein gel electrophoresis, mass spectrometry, spectroscopic analysis and functional assays.
Scale-Up production
If large quantities of the recombinant protein are required, the production process can be scaled up by culturing the transformed host cells in bioreactors. Bioreactors provide a controlled environment for the growth of cells and the production of recombinant proteins on a larger scale.
Protein expression systems
Protein expression systems involve different methods and organisms to produce recombinant proteins. The choice of expression system depends on factors such as the specific protein being produced, the desired protein characteristics, the required scale of production, the downstream applications and the available resources and expertise.
Protein expression systems include bacterial expression systems, yeast expression systems, insect cell expression systems, mammalian cell expression systems, cell-free expression systems and transgenic animals and plants. These factors play a crucial role in achieving high yields of functional recombinant proteins. Here is a brief overview of some commonly used protein expression systems:
  • Bacterial expression systems
Bacteria, such as Escherichia coli (E. coli), are widely used for recombinant protein expression due to their rapid growth, ease of cultivation and well-characterized genetics. Bacterial expression systems are generally cost-effective and offer high protein yields.
  • Yeast expression systems
Yeasts, such as Saccharomyces cerevisiae and Pichia pastoris, are commonly used for recombinant protein production. They offer advantages such as efficient protein folding, the ability to perform post-translational modifications, and scalability.
  • Insect cell expression systems
Insect cells, such as those derived from the fall armyworm, Spodoptera frugiperda (Sf9), or the cabbage looper, Trichoplusia ni (High Five), are utilized in the baculovirus expression system. This system allows for producing complex, properly folded proteins, including those requiring post-translational modifications.
  • Mammalian cell expression systems
Mammalian cells, including Chinese hamster ovary (CHO) cells and human embryonic kidney (HEK) cells, are favored for producing proteins that require authentic humanlike post-translational modifications or proper protein folding. Mammalian cell expression systems offer the advantage of producing recombinant proteins that closely resemble their native counterparts.
  • Cell-free expression systems
Cell-free protein expression systems utilize cell extracts, such as those derived from E. coli or wheat germ, to produce recombinant proteins in a test tube without needing intact cells. These systems offer rapid protein synthesis and allow for easy manipulation of reaction conditions. They are particularly useful for producing toxic or membrane proteins and high-throughput screening.
  • Transgenic animals and plants
Transgenic animals and plants can be genetically modified to produce recombinant proteins in their milk, eggs, blood or plant tissues. This approach offers the potential for large-scale protein production but may involve complex regulatory requirements and ethical considerations.
Types of recombinant proteins
Recombinant proteins hold immense value in advanced therapies, such as insulin for diabetes and targeted cancer treatments. In research, they help study protein structure while they create genetically modified crops with improved traits in agriculture.
Furthermore, they also serve as enzymes for biofuel and pharmaceutical production in industries. The proteins revolutionize multiple fields, offering better treatments, scientific insights, sustainable agriculture and innovative industrial processes.
The types of recombinant proteins include therapeutic proteins, industrial enzymes, diagnostic proteins, research tools, vaccines, biomaterials, bioengineering, biosensors and biocatalysts.
Therapeutic proteins
Therapeutic proteins are among the most well-known and widely used recombinant proteins. They include hormones, growth factors, antibodies, enzymes and clotting factors used to treat various diseases and medical conditions. Examples include insulin for diabetes, erythropoietin for anemia and monoclonal antibodies for cancer therapy.
Industrial enzymes
Recombinant proteins are crucial in producing industrial enzymes used in various industries. These enzymes offer improved efficiency, specificity, and stability compared to traditional enzymes derived from natural sources. Examples include amylases used in the food industry, proteases in detergent manufacturing and cellulases for biofuel production.
Diagnostic proteins
These proteins can be used as specific markers or antigens to detect the presence of certain diseases or infections. For instance, recombinant antigens are employed in immunoassays to diagnose HIV, hepatitis and other infectious diseases. Recombinant proteins are also used as controls and calibrators in laboratory tests.
Research tools
These proteins serve as essential tools for scientific research. They are used to investigate protein structure, function and interactions, providing insights into biological processes and disease mechanisms.
These are utilized in protein purification, crystallization, structural biology, enzymology and drug discovery. They serve as reagents in experiments and assays to study protein-protein interactions, protein-ligand binding and protein signalling pathways.
Vaccines
These proteins are employed in the development and production of vaccines. Certain vaccines are made by expressing recombinant viral or bacterial proteins that elicit an immune response without causing the actual disease. Examples include the recombinant hepatitis B surface antigen used in the hepatitis B vaccine and recombinant subunit vaccines for human papillomavirus (HPV) and influenza.
Biomaterials and bioengineering
The proteins are utilized in biomaterials and bioengineering applications. They can be engineered to possess properties such as biocompatibility, bioactivity and structural integrity. Recombinant proteins are employed in tissue engineering, regenerative medicine and developing scaffolds for cell culture and organ transplantation.
Biosensors and biocatalysts
These artificially produced proteins are also integrated into biosensors to detect specific molecules or analytes. These proteins can bind to the target molecule and generate a measurable signal, enabling sensitive and selective detection. Additionally, recombinant proteins are engineered as biocatalysts in biotechnological processes, converting substrates into desired products with high specificity and efficiency.
Applications of recombinant proteins
Recombinant proteins have a wide range of applications in biomedical research, revolutionizing drug discovery and development, diagnostics and immunotherapy. These applications have significantly impacted the fields of biotechnology and medicine, leading to advancements in various areas of healthcare.
Drug discovery and development
The proteins play a crucial role in drug discovery and development processes. They enable scientists to study the structure and function of target proteins involved in diseases, leading to the identification of potential drug targets. These proteins can be used in high-throughput screening assays to test the efficacy of various compounds against specific targets, accelerating the drug discovery process.
Moreover, recombinant proteins are essential for preclinical and clinical studies of potential drug candidates. They serve as therapeutic agents, such as recombinant antibodies or enzymes, in preclinical animal models to evaluate efficacy and safety. Recombinant proteins also play a vital role in clinical trials, where they are used to assess the therapeutic potential of new drugs and monitor treatment responses.
Diagnostics
The proteins have had a significant impact on medical diagnostics in human healthcare. They are widely used in various diagnostic tests, providing accurate and reliable results.
In immunoassays, recombinant proteins are used as antigens to detect specific antibodies in patient samples, aiding in the diagnosis of infectious diseases, autoimmune disorders and allergies. These proteins can also be used as standards or controls in these assays to ensure the accuracy of the test results.
Furthermore, recombinant proteins are essential in the production of vaccines. They are used as antigens to stimulate the immune system and induce the production of protective antibodies. Recombinant protein-based vaccines have successfully prevented diseases such as hepatitis B, HPV and influenza.
Additionally, recombinant proteins are utilized in biochemical analysis techniques. They can serve as molecular markers or probes to study specific proteins’ expression, localization and function in cells and tissues. It aids in the understanding of disease mechanisms and the development of targeted therapies.
Immunotherapy
Recombinant proteins have made significant contributions to the field of immunotherapy. They are used as therapeutic agents to modulate the immune system and treat various diseases, particularly cancer.
Recombinant antibodies, such as monoclonal antibodies, have revolutionized cancer treatment. They can be designed to target cancer cells, either directly killing them or blocking specific signalling pathways involved in tumor growth. Recombinant antibodies have shown remarkable success in treating several cancers, improving patient outcomes and survival rates.
Furthermore, recombinant proteins, such as cytokines and growth factors, are used in immunotherapy to enhance immune responses against tumors. They can stimulate the immune system, activate immune cells and promote anti-tumor immune reactions. These recombinant proteins have shown promise in boosting the efficacy of cancer immunotherapies, including immune checkpoint inhibitors and adoptive cell therapies.
Challenges in producing high-quality recombinant proteins
Producing high-quality recombinant proteins poses folding, stability, modifications, contamination, yield, solubility and scaling challenges. Proper folding ensures functionality, stability prevents degradation, modifications add complexity, contamination must be controlled, yield meets demand, solubility affects functionality and scaling requires optimization. As a result, addressing these challenges is important to obtain reliable and high-quality protein production.
Protein folding and stability
Recombinant proteins may face difficulties in correctly folding into their native three-dimensional structures. Improper folding can result in loss of protein function or aggregation. Optimizing the protein expression conditions, such as temperature, pH and co-expression of chaperones, is often necessary to improve protein folding and stability.
Post-translational modifications
Some proteins require post-translational modifications (PTMs) to achieve their functional form. However, certain expression systems may not possess the necessary cellular machinery for performing complex PTMs, such as glycosylation, phosphorylation or disulfide bond formation. These limitations can affect the functionality and efficacy of the recombinant protein.
Contamination and Impurities
Contamination with impurities, such as host cell proteins, nucleic acids, adventitious viruses and endotoxins, can affect the quality and safety of recombinant proteins. Strict purification processes are required to remove these contaminants and ensure the final product is pure and free from undesirable substances.
Low yield and productivity
Achieving high-protein yields is crucial for meeting the demand for recombinant proteins. However, low expression levels or inefficient protein production can lead to low yields. Factors such as codon usage, transcriptional regulation and protein degradation can impact protein productivity and require optimization.
Protein solubility and aggregation
Some recombinant proteins tend to aggregate, leading to loss of function and difficulties in purification. Strategies such as protein engineering, including adding fusion tags or optimizing expression conditions, may be necessary to improve protein solubility and prevent aggregation.
Scale-up challenges
Scaling up the production of recombinant proteins from laboratory scale to industrial scale can introduce additional challenges. Maintaining consistent protein quality, ensuring reproducibility and managing large-scale culture conditions and purification processes require careful optimization and validation.
Addressing these challenges requires a multidisciplinary approach involving protein engineering, optimization of expression systems, purification techniques and quality control measures. For further information on bioprocessing recombinant proteins and addressing these challenges, you can visit the following link for more resources and solutions by Avantor.