ContactUsLinkComponent
Downstream1.jpg
Downstream Processing Overview
Downstream processing is an essential aspect of biotechnology, particularly in the production of biopharmaceuticals. It includes the purification, isolation and characterization of the target product from complex biological matrices to ensure the final product is safe, effective and high-quality. However, downstream processing can be challenging due to factors such as the target product, concentration and environment.
This article will present an overview of downstream processing, including the steps, techniques and challenges. It will also cover the importance of downstream processing in biopharmaceutical production and the impact of recent technological advancements.
What is downstream processing?
Downstream processing involves the techniques and procedures to purify and recover a target product from a complex mixture of biological materials that include cells, proteins and nucleic acids. The target product is usually obtained through upstream processing, including the biological material’s growth and cultivation.
The goal of downstream processing is to isolate the target product from the mixture while also removing any contaminants that could interfere with its intended use. The process involves a series of steps: separation, purification, concentration and formulation. However, the specific techniques used in downstream processing may vary depending on the target product’s characteristics and the type of biological material from which it is derived.
There are four vital steps in downstream processing. They are:
  • Separation when the target product is separated from the mixture. Separation is often achieved through filtration, centrifugation or precipitation.
  • Purification, which involves removing contaminants such as other proteins, DNA or viruses. This step can be achieved through chromatography.
  • Concentration, in which the concentration of the target product is increased to the desired level.
  • Formulation, which includes preparing the product for its intended use by adding stabilizers or adjusting the pH level.
What are the steps in downstream processing?
The steps involved in downstream processing depend on the product type being produced and the manufacturing process. The general steps involved in downstream processing are as follows:
Harvesting and Clarification
The first step in downstream processing is harvesting and clarification. It involves the removal of unwanted impurities from the cell culture or biological source. Generally, different methods are employed to harvest the material depending on the type of biological source.
For instance, animal or plant tissue is mechanically disrupted or treated with enzymes to release the desired product. The microbial cultures for these tissues are harvested by centrifugation or filtration.
After harvesting, the material is subjected to clarification to remove unwanted impurities, such as cell debris, proteins and nucleic acids. It is achieved through filtration, centrifugation and precipitation techniques.
Separation
The second step in downstream processing is separation. In this step, the desired product is isolated from the other components in the harvested material. Several methods are employed for separation, including:
Centrifugation
Centrifugation uses centrifugal force to separate particles of different sizes and densities in a liquid mixture. This method is widely used to separate cells and organelles from cell cultures.
Filtration
Filtration is a process that passes a mixture through a filter to separate particles according to their size. Several types of filters can be used — including ultrafiltration, microfiltration or nanofiltration — depending on the size of the particles
Chromatography
Chromatography can be used to separate a mixture into its compounds by their differential absorption properties onto a stationary phase. The stationary phase can be a solid matrix or liquid.
Precipitation
This process separates compounds from a mixture by their differential solubility in a solvent. The desired product can be precipitated by adjusting the pH or mixture temperature, or by adding a precipitation agent, such as a salt or organic solvent.
Purification
Purification is a crucial step in the drug development process that involves removing impurities that may be present in the isolated product. The required purity level will depend on the product’s intended use, but in general, high purity is important to help ensure the safety and efficacy of the drug.
Several purification techniques are applicable, such as
  • Chromatography
  • Filtration
  • Distillation
  • Crystallization
Chromatography and filtration work the same during purification as they do in filtration. Distillation involves separating components of a mixture based on their boiling points and can often be used to help purify liquids. Crystallization is a process that forms high-quality crystals of the desired product from liquids by controlling the cooling rate and evaporating solvents
Formulation
Formulation is the stage in which drugs are placed into the form needed to deliver a specific drug dose to patients. Drugs should be formulated to help ensure their stability and efficacy. The process involves adding excipients, or non-active ingredients, to achieve those properties.
Excipients can help improve solubility, stability or bioavailability. For example, stabilizers such as sugars or amino acids can be added to protect against drug degradation, while buffers can help maintain a consistent pH. Preservatives may be added to help prevent microbial growth.
The formulation step will depend on the drug’s intended use. Drugs administered orally, for example, may be formulated as tablets, capsules or liquids, while drugs that are swallowed as liquids may be formulated as a solution or a suspension.
Final Product Filling and Packaging
The final step in downstream processing involves filling products into their final containers and packaging the containers for distribution. This critical step may determine the product’s shelf life and stability. It’s essential that filling is completed under strict aseptic conditions to help prevent microbial contamination.
In addition, filling equipment must be validated and calibrated to ensure accurate volumes. After filling, products are sealed with the correct closure systems to prevent contamination, and labeling is added to provide critical information such as product name, dosage and expiration date.
Downstream processing development
Downstream processing covers various methods and techniques to separate and purify biomolecules, including proteins, antibodies, enzymes and nucleic acids. The downstream processing development strategies aim to optimize the purification process’s efficiency, yield, purity and cost-effectiveness.
The methods used in downstream processing can be categorized into several key areas, each focusing on specific aspects of purification and separation. These methods include:
Cell Harvesting
Cell harvesting is the initial step in downstream processing, where cells or microorganisms containing the desired product are separated from the culture medium. Centrifugation, microfiltration and flocculation are commonly employed techniques for cell harvesting
Centrifugation separates cells based on their size and density, while microfiltration employs porous membranes to retain cells and allow the passage of the culture medium. Flocculation requires adding flocculating agents to aggregate cells and facilitate the separation process.
Cell Disruption
Cell disruption is performed to release intracellular components such as proteins, enzymes and nucleic acids from the harvested cells. Bead milling, high-pressure homogenization and ultrasonication are mechanical methods to break open the cells and release their contents. Enzymatic methods utilizing specific enzymes to degrade the cell wall can also be employed.
Filtration
Filtration is a widely used technique in downstream processing to separate particulate matter, impurities and cellular debris from the desired product. This step can involve various filtration methods, including depth, membrane filtration (microfiltration, ultrafiltration, nanofiltration) and chromatography.
Chromatography
Chromatography plays a significant role in downstream processing for the separation and purification of biomolecules. Different chromatographic techniques, such as affinity chromatography, ion exchange chromatography, size exclusion chromatography and hydrophobic interaction chromatography, are employed based on the physicochemical properties of the target molecule. Chromatography utilizes stationary and mobile phases to separate and purify the desired product from impurities.
Precipitation
Precipitation Precipitation involves the addition of specific agents to induce the formation of a solid precipitate, which can be separated from the solution by centrifugation or filtration. This method is useful for the removal of impurities or for concentrating the desired product. Various techniques, like salting out, pH precipitation and temperature-induced precipitation, can be employed.
Crystallization
Crystallization is employed to purify the solid products or to produce highly purified substances. By controlling factors such as temperature, concentration and pH, the desired product can be induced to form crystals while impurities remain in the solution or are excluded from the crystal lattice. The crystals can then be separated and further purified.
Drying
Drying is the final step in downstream processing, where the purified product is dried to remove residual water and obtain a stable solid or powder form. Common drying techniques include freeze-drying (lyophilization), spray drying and vacuum drying. These methods help to preserve the stability and extend the shelf life of the purified product.
The development of downstream processing methods involves a combination of experimentation, process optimization and innovation to achieve high yields, purity and cost-effectiveness. Factors such as product characteristics, scalability and regulatory requirements also play a crucial role in determining the most suitable methods for a specific application.
Why is downstream processing important?
Downstream processing is a critical aspect of biotechnology because it enables the separation, purification and recovery of a target product from a complex mixture of biological materials. The target product can be a protein, nucleic acid, virus or cell used in various applications, including biopharmaceuticals, industrial enzymes and vaccines.
Downstream processing involves a series of complex steps that can be time-consuming and costly. The purification of a target product often requires multiple steps of chromatography, filtration and centrifugation, which can take several days to complete. Additionally, the equipment and materials needed for downstream processing can be expensive and require specialized knowledge and skills.
Another significant pain point in downstream processing is the potential for product degradation. The target product can be easily denatured, aggregated or degraded by environmental factors such as pH, temperature and shear forces. It can lead to the loss of product yield and quality loss, significantly impacting the final product’s safety and efficacy.
Despite these challenges, downstream processing is essential for producing safe and effective biopharmaceuticals. The purity and quality of the final product are critical for regulatory approval and patient safety. Technological advancements, such as high-throughput screening and automated purification systems, have recently improved the downstream processing’s efficiency and speed, making it more accessible and cost-effective for the biotech industry.
Considerations for a successful downstream bioprocessing
Successful downstream bioprocessing is important to purify and isolate valuable products from biological sources such as cells, tissues or fermentation broths. This stage of bioprocessing follows upstream activities like cell culture or fermentation and involves steps such as cell harvesting, clarification, filtration, purification and formulation. Here are some key considerations to ensure a successful downstream bioprocessing:
Process Understanding
Thoroughly understand the characteristics of the biological material and the desired product. This includes knowledge about the target molecule, such as its stability, solubility and susceptibility to degradation or aggregation. Understanding the composition of the feedstock and potential impurities is crucial for designing effective purification strategies.
Robust Harvesting and Clarification
Develop efficient methods for cell harvesting or broth clarification to remove solids, cell debris and impurities. Optimization of centrifugation, filtration or flocculation techniques is essential to maximize yield and minimize product loss.
Scalability
Consider scalability throughout the downstream process. Ensure that the chosen purification techniques and equipment can handle increasing volumes as production scales up. Pilot-scale testing and process validation are vital to anticipate challenges and optimize the process for large-scale production.
Purification Techniques
Select appropriate purification techniques based on the characteristics of the target molecule. Common techniques include chromatography (affinity, ion exchange, size exclusion), filtration (microfiltration, ultrafiltration) and precipitation. Evaluate different options and develop a purification strategy that provides high product yield, purity and activity.
Optimization of Parameters
Optimize critical process parameters such as temperature, pH, pressure and flow rates to maximize product recovery and quality. Conduct robust design-of-experiment (DoE) studies to identify optimal conditions and ranges for each step of downstream processing.
Product Stability
Ensure product stability during purification and formulation steps. Implement strategies to prevent degradation, denaturation or aggregation of the target molecule. Consider using appropriate buffers, stabilizers and formulation additives to maintain product integrity and activity.
Quality Control and Analytics 
Implement a comprehensive analytical strategy to monitor product quality at different stages of downstream processing. Utilize techniques like chromatography, electrophoresis, spectroscopy and bioassays to assess purity, identity, potency and safety of the product. Implement stringent quality control measures to ensure consistency and batch-to-batch reproducibility.
Regulatory Compliance
Stay up to date with regulatory requirements and guidelines for downstream bioprocessing. Ensure compliance with Good Manufacturing Practices (GMP) and other relevant regulations to ensure product safety, quality and traceability. Document all process steps and maintain comprehensive records for audits and regulatory submissions
Takeaways:
  • Understand the biological material’s and target molecule’s characteristics to design an effective purification strategy
  • Optimize critical process parameters and evaluate scalability for large-scale production
  • Select appropriate purification techniques based on the target molecule’s properties
  • Ensure product stability and prevent degradation or aggregation during processing
  • Implement comprehensive quality control measures and stay compliant with regulatory requirements
By considering these factors, bioprocessing scientists can enhance the efficiency and success of downstream processing, leading to the production of high-quality biotherapeutics or valuable biomolecules.
Reduce downstream turnaround times and optimize buffer and chromatography resin selection in biopharma mAbs manufacturing with Avantor. Learn about our downstream processing services and how our experts can help.
Let us help you refine your discoveries and achieve your scientific objectives. Visit Avantor today to benefit from the groundbreaking scientific advancements. Our innovative products and services are designed to meet your laboratory requirements, from chemicals and reagents to lab equipment and clean room supplies.