Orders LinkContactUsLinkComponent
Accredited Thermometer Calibration and Repair Service
Accredited Thermometer Calibration and Repair Service
Brand-independent, accurate, competence....
Accredited Thermometer Calibration and Repair Service​ | Avantor
Our ISO17025-accredited calibration-facility is able to calibrate and adjust all brands of temperature-devices like thermometers, dataloggers, blocks and baths.
Your advantages:
  • Approved quality
  • Detailed Proces-documentation
  • All brands & models
  • NIST traceable
  • ISO 17025 calibration-certificate
  • Due time reminder system
  • Combine with humidity-calibration
Download Order/Decontamination Form
calibration_400x495.png
For temperature measurement our laboratory, can calibrate all types of dial type temperature indicators, dataloggers, recorders, liquid-in-glass thermometers and temperature sensors. A fast, efficient and competitive service is provided to examine, report on and calibrate customers’ instruments in addition to supplying new, calibrated thermometers and dataloggers. Before calibration, all instruments are examined and checked to confirm they are free from any defects. Customers are automatically advised of any non-conformities of instruments they have submitted, prior to calibration.
Request for quotation
ThermometerCalibration.png
  • In-house calibration facility in Bristol
  • UKAS calibration of total and partial immersion liquid in glass thermometers between -35 °C and 650 °C
  • UKAS calibration of digital thermometers (system calibrations) between -35 °C and 650 °C
  • Best measurement capability of 0.024 oc using our high specification calibration equipment
Download pdf