Orders LinkContactUsLinkComponent
thermometer-calibration-feature.jpg
Accredited Thermometer Recertification & Calibration Services
Avantor Services offers recertification, calibration, and repair services for all makes and models of thermometers.
Request a Quote Today
Thermometer Calibration | Avantor
Our thermometer services include:
  • Complete visual inspection for any physical damage. If possible, the damage will be repaired.
  • Recertification from 1 Point to 15 Points
  • Thermometer readings are compared against NIST standards in high precision calibration baths, using the comparison method at determined test temperatures
  • Each temperature is recorded to 1/10th of a scale division
  • A calibration report is generated, detailing pertinent information including the correction factors for each test point
  • All calibration information is entered into a data storage system
  • All certification procedures are performed in accordance with NIST and ASTM guidelines