Orders LinkContactUsLinkComponent
Balances | Avantor
Balances
Whether you need to weigh very accurately in a lab using complex programs, weigh recipes in production, use your scale for counting purposes or simply weigh at school or in your kitchen, we offer a scale for every application.
From micro balances with a readability d of 1 µg, semi micro balances, analytical balances with readability d of 0,1 mg, to precision balances, in different quality levels depending on your applications.
Balances with a readability ‘d’ of 100 mg (0,1 g) or higher and high weighing range.
A wide range of reliable moisture balances to meet the needs of both quality and in-process control.
Featured content
Equipment-AndInstrument.png
Optimizing uptime and minimizing downtime.
weighing-balance_new.png
All the tips, tricks, consumables and equipment you need in a weighing workflow.
Let us help you move science forward.