ContactUsLinkComponent
UVVIS-spectroscopy.jpg
UV/VIS spectroscopy
UV/Vis analysis with a spectrophotometer can require different types of chemicals. First, a specific grade of solvents is developed to avoid optical impurities of your sample. Second, the spectrophotometer needs to be calibrated using UV/Vis standard kits. Third, color reference solution kits can save you time in preparing the reference solutions yourself.
VWRCHEMICALS.jpg
High purity solvents specially designed for spectroscopy applications that demand the highest spectral purity.
UVVIS-Standards.jpg
Be sure of your results with our complete range to calibrate spectrophotometers on the control of absorbance, resolution power, stray light and wavelength accuracy
css.jpg
In petrochemistry mainly, we can offer complete range of "ready to use" solutions for use as calibration and/or quality control standards according to ASTM, Saybolt, Platinum-coblat and European Pharmacopoeia (Ph Eur)
Colour-reference.jpg
These solutions are developed and produced according to chapter 2.2.2. "Degree of coloration of liquids" of the European Pharmacopeia.
Frequently Asked Questions
What kind of standards are available to calibrate my UV/Visible spectrophotometer?
Different standards in ampoules delivered in kits with a complete Certificate of Analysis are available to check parameters such as:
  • Absorbance control
  • Stray light testing
  • Resolution power testing
  • Wavelength testing
Additional Resource