ContactUsLinkComponent
Chemicals for Sample Preperation
Sample preparation is as important as the analysis of your sample. Make sure you use the right reagents to avoid contamination of your sample. Avantor® carries everything you need.
Are inorganic salts BSE-free?
All inorganic salts are produced from inorganic raw materials. Since no animal-derived material is used in the production of these compounds it can be assumed that they are BSE-free.
How do decahydrates and dodecahydrates have to be stored?
Decahydrates and dodecahydrates such as sodium carbonate decahydrate, sodium sulfate or di-sodium hydrogen phosphate dodecahydrate lose some their crystallization in water at 33 °C. It can be said that they melt in their own crystallization water. It is, therefore, important to store and transport these compounds under cool conditions (below 20 °C) mainly during the summer months.
How about the shelf life of salts?
Some salts such iron (II) sulfate heptahydrate, ammonium iron (II) sulfate hexahydrate or tin (II) chloride dihydrate are potent reducing agents, which at higher temperatures or when exposed to direct sunlight, readily change into a higher oxidation state. These products must, therefore, be stored under cool, dark conditions (15 °C). Nevertheless, the minimum shelf life of these products is limited to 1 year, even under ideal storage conditions.
What are stabilized products
Some compounds are explosive and may not be sold as dry substances but only mixed with water. For instance, ammonium dichromate (stab. with 0.5% to 3% water), ammonium nitrate (min. 3% water ) or 2,4 dinitrophenol (0,5 ml H2O2/g).
How can iodine be dissolved in water?
Iodine is only slightly soluble in water, whereas it readily dissolves in a concentrated potassium iodide solution by forming complexes. The solution should, however, be diluted further only after all the iodine dissolves. This is very slow despite the presence of potassium iodide.
Are pack sizes available for inorganic salts?
There are many different pack sizes available such as 100, 250, 500, 2500 or 5000 g. For some products 25 kg is also available. Check out our website or chemical catalogs.
Do inorganic lubricants exist?
Besides frequently used organic lubricants, there are also inorganic lubricants. The lubricating effect is based on the layered structure of these compounds. Common lubricants consist of boron nitride, molybdenum (IV) sulfide or graphite fine powder. These compounds can be used both at low and higher temperatures.
What are vitriols?
Vitriols are sulfates or divalent metals in their hydrated, crystalline form such as copper (CuSO4. 5H2O), iron (FeSO4.7H2O) or zinc (ZnSO4.7H2O).
What is the difference between ammonium carbamate and ammonium carbonate?
Ammonium carbonate is the salt of carbonic acid and ammonium carbamate is the salt of carbamic acid. Ammonium carbamate is much more stable than ammonium carbonate and changes into the latter only in aqueous solution as is the case when boiling it.
What are caustic alkalis and alkali solutions?
Caustic alkali is a term used to call the hydroxides of alkali metals. They occur in solid form and are generally manufactured as pellets. Alkali solutions are the aqueous versions of these hydroxides. The most commonly used caustic alkalis and alkali solutions are those of sodium and potassium.
How can alkali solutions be assayed?
Alkali solutions can be directly titrated with standard acids.
What safety precautions should you take when mixing alkali solutions with acids?
If possible, and only if necessary, mix nothing other than dilute alkali solutions with dilute acids whilst stirring vigorously. Wear protective gloves, glasses and overalls, and work under a hood. Very cautiously stir the alkali solution into the acid. Pay careful attention to all rules for safe working.
What is the best way of dissolving pellets?
Slowly and vigorously stir the required quantity of sodium hydroxide or potassium hydroxide pellets into water at +20 °C. Be aware that a great deal of heat may be produced.
What is the procedure for disposing alkali solutions.
When necessary, these solutions can be diluted by cautiously stirring into water, followed by neutralization with hydrochloric acid. Be sure to wear protective gloves and glasses and to work under a hood. Before emptying into a collection container, make sure you take a pH reading using pH universal indicators strips.
Are sodium hydroxide and Potassium hydroxide pellets soluble in organic solvents?
The pellets are soluble in alcohol and insoluble in acetone and diethyl ether.
What are monobasic and dibasic phosphates?
Disodium hydrogen phosphate (Na2HPO4) is designated a dibasic phosphate and sodium dihydrogen phosphate (NH2PO4) a mono basic phosphate.
Why do more concentrated solutions have a lower carbonate content?
Carbonates are only slightly soluble in sodium and potassium hydroxide solutions. The more concentrated the solution is, the lower the carbonate content, as carbonates precipitate out.
Complete range of solvents suitable for a multitude of analytical and R&D applications with guaranteed specifications conform to ACS and Reag PH EUR for most of them, available in different pack size.
For sample preparation, PH adjusters or other applications, acids and alkalis are often used. You will find a complete range of these products in different packs.
Complete range of salts in analytical grade conform to ACS, Reag PH EUR for the big
For quality control in pharmaceutical companies, over 250 standards and solutions conform to the last PH EUR version delivered with a complete CoA insuring a complete traceability.
Solvents and acids in plastic bottles are safe, virtually unbreakable, environmentally, friendly, light and easy to handle.
This kind of "safe" packaging is really helpful for corrosive reagents. The PE-coating contains the product when the glass bottle is broken.