Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Qualitative Lead acetate paper for Sulfide: 5 mg/L S²⁻, (booklet) 1 * 100 items
Catalog # VWRC92524.601
Supplier:
VWR Chemicals
New Product
Expand image
Qualitative Lead acetate paper for Sulfide: 5 mg/L S²⁻, (booklet) 1 * 100 items
Catalog # VWRC92524.601
Supplier:
VWR Chemicals
New Product