Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
BUFFER STANDARD PH 12.00 A 25°C ISO17034 1 * 1 L
Catalog # REGECRM112025
Supplier:
REAGECON
New Product
Expand image
BUFFER STANDARD PH 12.00 A 25°C ISO17034 1 * 1 L
Catalog # REGECRM112025
Supplier:
REAGECON
New Product