Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Includes 5x Proteometer-CV Fluorescent Reagent Vials. Allows for analysis up to 300 samples - PLEASE ADD STARTER PROT-CV KIT SEPERAT LEY (KIT-PCV-003-01A) 1 * 1 KIT
Catalog # NOVIKIT-PCV-00301B
Supplier:
NOVILYTIC
New Product
Expand image
Includes 5x Proteometer-CV Fluorescent Reagent Vials. Allows for analysis up to 300 samples - PLEASE ADD STARTER PROT-CV KIT SEPERAT LEY (KIT-PCV-003-01A) 1 * 1 KIT
Catalog # NOVIKIT-PCV-00301B
Supplier:
NOVILYTIC
New Product