ContactUsLinkComponent
Includes 1x Mixing Tee, 2x Tubing Unions, 16x Prot-CV MD Buffer Packets, 20x Prot-CV MPA Salt Packets, 4x Prot-CV MPB Salt Packets, 1x Reconstitution Reagent - Allows for analysis up to 240 samples - ADD PROT-CV FLUORESCENT REAGENT SEPERATLEY (4x RGN 1 * 1 KIT
Catalog # NOVIKIT-PCV-005-01
Supplier:  NOVILYTIC
New Product
Includes 1x Mixing Tee, 2x Tubing Unions, 16x Prot-CV MD Buffer Packets, 20x Prot-CV MPA Salt Packets, 4x Prot-CV MPB Salt Packets, 1x Reconstitution Reagent - Allows for analysis up to 240 samples - ADD PROT-CV FLUORESCENT REAGENT SEPERATLEY (4x RGN 1 * 1 KIT
Catalog # NOVIKIT-PCV-005-01
Supplier:  NOVILYTIC
New Product