Proteometer-CV Fluorescent Reagent
NOVIRGN-PCV-001
New Product
:
- Description:Proteometer-CV fluorescent reagent. 1 vial of fluorescent reagent for the Proteometer-CV kit
Replacement fluorescent kit reagent for the Proteometer-CV.
- Replacement vials for frequent users of the Proteometer-CV kit
- Store at −20 °C
Intended for use with the Proteometer-CV kit. Crimped, sealed, sold individually.
: Supplied in a custom box with instructions.