Proteometer-CV Full Kit (300 Assays)
Catalog # NOVIKIT-PCV-00301A
Supplier: NOVILYTIC
New Product
Specifications
- Description:Proteometer-CV full kit (300)
Specifications
About this item
The Proteometer-CV full kit provides 300 assays for charge variant analysis. It runs in 25 minutes or less.
- No sample preparation required - analysis directly from clarified fermentation broth without Protein A purification
- Speed and efficiency - CV analysis in under 25 minutes
- Simplified workflow - all the components you need for seamless operation
- Exclusive technology - the only solution that performs charge variant analysis directly in clarified fermentation broth
The Proteometer-CV kit offers an alternative to traditional charge variant methods that require Protein A purification, thereby saving time and money. We are the only solution that performs charge variant analysis of samples directly in clarified fermentation broth, minimising sample handling required for other charge variant analyses.
Accessories information: See the Proteometer-CV reactor, Proteometer-CV buffer packets, and Proteometer-CV reagent series for accessory component replacement options.
Ordering information: Glassware not included. Please add Proteometer-CV fluorescent reagent seperately.
Delivery: Includes 1× Proteometer-CV reactor, 1× MD unit, 1× mixing tee, 1× tubing unions, 1× tubing connector, 20× Prot-CV MD buffer packets, 25× Prot-CV MPA salt packets, 4× Prot-CV MPB salt packets, 1× reconstitution reagent - allows for analysis up to 300 samples.
Packaging: Supplied in a custom box with instructions and how-to video. No refrigeration or special handling required.