ContactUsLinkComponent
Includes 1x Proteometer-CV Reactor, 1x MD Unit, 1x Mixing Tee, 2x Tubing Unions, 4x Prot-CV MD Buffer Packets, 5x Prot-CV MPA Salt P ackets, 1x Prot-CV MPB Salt Packets, 1x Reconstitution Reagent - Allows for analysis up to 60 samples - ADD PROT-CV F 1 * 1 KIT
Catalog # NOVIKIT-PCV-00101A
Supplier:  NOVILYTIC
New Product
Includes 1x Proteometer-CV Reactor, 1x MD Unit, 1x Mixing Tee, 2x Tubing Unions, 4x Prot-CV MD Buffer Packets, 5x Prot-CV MPA Salt P ackets, 1x Prot-CV MPB Salt Packets, 1x Reconstitution Reagent - Allows for analysis up to 60 samples - ADD PROT-CV F 1 * 1 KIT
Catalog # NOVIKIT-PCV-00101A
Supplier:  NOVILYTIC
New Product