1 component: Cyclohexane [CAS:110-82-7] 0.4g/ l in Methanol CRM, ISO 17034 and ISO 17025 Conservation 12 months. Store in a refrigerator at temperatures between 2°C to 8°C packaging: 5*1ml ampoule 1 * 5 Ampoul
Catalog # CMPLRD1223031.5
Supplier: CUSTOM MADE CHEMICALS LAB
New Product