Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Muffle Furnace LT5/13/B510 Tmax1300 with lift door and over-temperature limiter and thermocouple port in the rear wall 1 * 1 items
Catalog # 455-3188
Supplier:
NABERTHERM
New Product
Expand image
Muffle Furnace LT5/13/B510 Tmax1300 with lift door and over-temperature limiter and thermocouple port in the rear wall 1 * 1 items
Catalog # 455-3188
Supplier:
NABERTHERM
New Product