Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Muffle Furnace L5/13/C550 Tmax1300 with flap door and thermocouple hole in the furnace door 1 * 1 items
Catalog # 455-3159
Supplier:
NABERTHERM
New Product
Expand image
Muffle Furnace L5/13/C550 Tmax1300 with flap door and thermocouple hole in the furnace door 1 * 1 items
Catalog # 455-3159
Supplier:
NABERTHERM
New Product