Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Muffle Furnace L5/13/C550 Tmax1300 with flap door and over-temperature limiter and thermocouple hole in the furnace door 1 * 1 items
455-3143
:
NABERTHERM
New Product
Muffle Furnace L5/13/C550 Tmax1300 with flap door and over-temperature limiter and thermocouple hole in the furnace door 1 * 1 items
455-3143
:
NABERTHERM
New Product