ContactUsLinkComponent
Muffle Furnace L40/12/C550 Tmax1200 with flap door and protective gas connection, over-temperature limiter and thermocouple port in the rear wall 1 * 1 items
Catalog # 455-3066
Supplier:  NABERTHERM
New Product
Muffle Furnace L40/12/C550 Tmax1200 with flap door and protective gas connection, over-temperature limiter and thermocouple port in the rear wall 1 * 1 items
Catalog # 455-3066
Supplier:  NABERTHERM
New Product