ContactUsLinkComponent
Muffle Furnace LT40/11/B510 Tmax1100 with lift door and protective gas connection, over-temperature limiter and thermocouple port in the rear wall 1 * 1 items
Catalog # 455-3030
Supplier:  NABERTHERM
New Product
Muffle Furnace LT40/11/B510 Tmax1100 with lift door and protective gas connection, over-temperature limiter and thermocouple port in the rear wall 1 * 1 items
Catalog # 455-3030
Supplier:  NABERTHERM
New Product