ContactUsLinkComponent
Muffle Furnace LT24/12/C550 Tmax1200 with lift door and protective gas connection, over-temperature limiter and thermocouple port in the rear wall 1 * 1 items
  455-2994
New Product
Muffle Furnace LT24/12/C550 Tmax1200 with lift door and protective gas connection, over-temperature limiter and thermocouple port in the rear wall 1 * 1 items
  455-2994
New Product