ContactUsLinkComponent
Muffle Furnace L15/12/C550 Tmax1200 with flap door and protective gas connection, over-temperature limiter and thermocouple hole in furnace door 1 * 1 items
  455-2875
New Product
Muffle Furnace L15/12/C550 Tmax1200 with flap door and protective gas connection, over-temperature limiter and thermocouple hole in furnace door 1 * 1 items
  455-2875
New Product