Order Entry
Export
Orders LinkContactUsLinkComponent
Mouse Recombinant FGF-15 (from E. coli)

Mouse Recombinant FGF-15 (from E. coli)

Catalog # ABCAAB315426-1MG
New Product
Supplier:   Abcam
Mouse Recombinant FGF-15 (from E. coli)
Catalog # ABCAAB315426-1MG
Supplier:  Abcam
New Product
CAS Number:  
Mouse Recombinant FGF-15 (from E. coli)
Mouse Recombinant FGF-15 (from E. coli)
CAS Number:  
Price
3 150,00 €
Log in for availability
1 * 1 mg
Product Details & Documents

Recombinant Mouse FGF-15 protein is a Mouse Full Length protein, in the 26 to 218 aa range, expressed in E. coli, with >90% purity and suitable for SDS-PAGE.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications

Specifications

  • Form:
    Lyophilised
  • Protein/peptide name:
    FGF-15
  • Protein synonyms:
    Fibroblast growth factor 15|FGF-15|Fgf15
  • Purity:
    >90%
  • Source:
    E. coli
  • Shipping temperature:
    20
  • Cat. No.:
    ABCAAB315426-1MG
  • Protein/peptide type:
    Recombinant
  • Storage conditions:
    Store at −20 °C
  • Formulation:
    pH: 7,4 - 8. Constituents: 6% Trehalose, 0,87% Ethylenediaminetetraacetic acid, disodium, dihydrate, 0,24% Tris, 0,05% dodecyl 2-(trimethylazaniumyl)ethyl phosphate.
  • Conjugation:
    Unconjugated
  • Tested applications:
    SDS-PAGE
  • Species:
    Mouse
  • Animal-free:
    N

Specifications