Human Elastin ELISA Kit
Human Elastin ELISA kit, 384 tests
ABCAAB239433-1X384
New Product
: Abcam
Human Elastin ELISA Kit is a single-wash 90-min Simplestep used to quantify Human Elastin with a sensitivity of 44 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 384-well plate for higher throughput.
- Description:Human Elastin ELISA kit
- Tests per kit:384 tests
- Primary antibody reactivity:Human
- Sample type:Citrate plasma, Serum, EDTA Plasma
- Detection range:0,19 - 12 ng/ml
- Storage temperature:+4 °C
- Assay Type:Sandwich
- Time to Results:1 h 30 min
- Cat. No.:ABCAAB239433-1X384
- Assay duration:One step
- Detection method:Colorimetric
- Sensitivity:44 pg/ml
- Format:Pre-coated
- Target protein:Elastin
- Regulatory status:RUO



