Mouse Beta-2-Microglobulin ELISA Kit
Mouse Beta-2-Microglobulin ELISA Kit, 384 tests
Catalog # ABCAAB223590-1X384
New Product
Supplier: Abcam
Mouse Beta-2-Microglobulin ELISA Kit
Catalog # ABCAAB223590-1X384
Supplier: Abcam
New Product
CAS Number:
Jump to:
Product Details & Documents
Mouse Beta-2-Microglobulin ELISA kit is a single-wash 90-min Simplestep used to quantify Mouse Beta-2-Microglobulin with a sensitivity of 1.165 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader. Different formats for different needs: 384-well plate for higher throughput.
Specifications
Specifications
- Description:Mouse Beta-2-Microglobulin ELISA Kit
- Tests per kit:384 tests
- Primary antibody reactivity:Mouse
- Sample type:Urine, Heparin Plasma, Citrate plasma, Cell culture supernatant, Serum, EDTA Plasma
- Detection range:3,125 - 200 pg/ml
- Storage temperature:4 °C
- Assay Type:Sandwich
- Time to Results:1 hr 30 min
- Cat. No.:ABCAAB223590-1X384
- Assay duration:One step
- Detection method:Colorimetric
- Sensitivity:1,165 pg/ml
- Format:Pre-coated
- Target protein:beta 2 Microglobulin
- Regulatory status:RUO



