ContactUsLinkComponent
7 replacement vials for the Proteometer-L Kit. Crimped, sealed, for when a user runs out but does not need to reorder a reactor and/or reagents. Intended for use with the Proteometer-L Kit 1 * 1 items
Catalog # NOVIRGN-PL-0017
Supplier:  NOVILYTIC
New Product
7 replacement vials for the Proteometer-L Kit. Crimped, sealed, for when a user runs out but does not need to reorder a reactor and/or reagents. Intended for use with the Proteometer-L Kit 1 * 1 items
Catalog # NOVIRGN-PL-0017
Supplier:  NOVILYTIC
New Product