ContactUsLinkComponent
The kit includes the Proteometer Reactor, 14 reagent vials, 14 buffer packets, and other parts needed for a plug and play use with any liquid chromatograph that has a fluoresence detector. PLEASE ADD REAGENT SEPARATELY Eliminate sample prep and m 1 * 1 SET
Catalog # NOVIKIT-PL-007-01A
Supplier:  NOVILYTIC
New Product
The kit includes the Proteometer Reactor, 14 reagent vials, 14 buffer packets, and other parts needed for a plug and play use with any liquid chromatograph that has a fluoresence detector. PLEASE ADD REAGENT SEPARATELY Eliminate sample prep and m 1 * 1 SET
Catalog # NOVIKIT-PL-007-01A
Supplier:  NOVILYTIC
New Product