Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
KÜCHENSCHAUFEL 0,6L, Edelstahl 18/10 Drm. 9,5cm, Länge 26,5cm (Schaufellänge 14cm) 1 * 1 ST
Catalog # CONC1036/090
Supplier:
CONTACTO
New Product
Expand image
KÜCHENSCHAUFEL 0,6L, Edelstahl 18/10 Drm. 9,5cm, Länge 26,5cm (Schaufellänge 14cm) 1 * 1 ST
Catalog # CONC1036/090
Supplier:
CONTACTO
New Product