ContactUsLinkComponent
Mouse Caspase-7 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinat ion of mouse Caspase-7 in tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA2486-96
Supplier:  Antibodies.com
New Product
Mouse Caspase-7 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinat ion of mouse Caspase-7 in tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA2486-96
Supplier:  Antibodies.com
New Product