ContactUsLinkComponent
Human Fascin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Fascin in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA2789-96
Supplier:  Antibodies.com
New Product
Human Fascin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Fascin in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA2789-96
Supplier:  Antibodies.com
New Product