ContactUsLinkComponent
Rat CXCR3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat CXCR3 in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
  ANTIA1840-96
New Product
Rat CXCR3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat CXCR3 in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
  ANTIA1840-96
New Product