0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Recombinant E. coli DNA Photolyase Protein (Functional) 1 * 20 µG
ANTIA121189-20
:
Antibodies.com
New Product
Recombinant E. coli DNA Photolyase Protein (Functional) 1 * 20 µG
ANTIA121189-20
:
Antibodies.com
New Product