ContactUsLinkComponent
Human EBI3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human EBI3 in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
  ANTIA270652-96
New Product
Human EBI3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human EBI3 in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
  ANTIA270652-96
New Product