ContactUsLinkComponent
Human EG-VEGF ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of human EG-VEGF in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological fl 1 * 96 Tests
Catalog # ANTIA1677-96
Supplier:  Antibodies.com
New Product
Human EG-VEGF ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of human EG-VEGF in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological fl 1 * 96 Tests
Catalog # ANTIA1677-96
Supplier:  Antibodies.com
New Product