ContactUsLinkComponent
Rat Pyruvate Kinase Isozyme M1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantita tive determination of rat Pyruvate Kinase Isozyme M1 in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA1830-96
Supplier:  Antibodies.com
New Product
Rat Pyruvate Kinase Isozyme M1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantita tive determination of rat Pyruvate Kinase Isozyme M1 in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA1830-96
Supplier:  Antibodies.com
New Product