ContactUsLinkComponent
Human pan IFN alpha Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibody, Streptavidin-AP, recombinant human IFN alpha ELISA standard, and standard reconstitution buffer. This matc 1 * 1.920 Tests
Catalog # ANTIA270417-2096
Supplier:  ANTIBODIES.COM
New Product
Human pan IFN alpha Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibody, Streptavidin-AP, recombinant human IFN alpha ELISA standard, and standard reconstitution buffer. This matc 1 * 1.920 Tests
Catalog # ANTIA270417-2096
Supplier:  ANTIBODIES.COM
New Product