ContactUsLinkComponent
Cotton Rat Interferon gamma Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal d etection antibody, Streptavidin-HRP, recombinant cotton rat Interferon gamma ELISA standard, and standard reconstitut 1 * 576 Tests
  ANTIA270465-696
New Product
Cotton Rat Interferon gamma Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal d etection antibody, Streptavidin-HRP, recombinant cotton rat Interferon gamma ELISA standard, and standard reconstitut 1 * 576 Tests
  ANTIA270465-696
New Product