ContactUsLinkComponent
Rabbit Lactate Dehydrogenase B/LDH-B ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qu antitative determination of rabbit Lactate Dehydrogenase B/LDH-B in serum, plasma, tissue homogenates, and other biol 1 * 96 Tests
Catalog # ANTIA303850-96
Supplier:  ANTIBODIES.COM
New Product
Rabbit Lactate Dehydrogenase B/LDH-B ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qu antitative determination of rabbit Lactate Dehydrogenase B/LDH-B in serum, plasma, tissue homogenates, and other biol 1 * 96 Tests
Catalog # ANTIA303850-96
Supplier:  ANTIBODIES.COM
New Product