ContactUsLinkComponent
Human Fibrinogen Degradation Product ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qu antitative determination of human Fibrinogen Degradation Product in serum, plasma, cell culture supernatant, cell or 1 * 96 Tests
Catalog # ANTIA326561-96
Supplier:  ANTIBODIES.COM
New Product
Human Fibrinogen Degradation Product ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qu antitative determination of human Fibrinogen Degradation Product in serum, plasma, cell culture supernatant, cell or 1 * 96 Tests
Catalog # ANTIA326561-96
Supplier:  ANTIBODIES.COM
New Product