ContactUsLinkComponent
Porcine IL-5 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibo dy, Streptavidin-HRP, recombinant porcine IL-5 ELISA standard, and standard reconstitution buffer. This matched antib 1 * 576 Tests
Catalog # ANTIA325356-696
Supplier:  ANTIBODIES.COM
New Product
Porcine IL-5 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibo dy, Streptavidin-HRP, recombinant porcine IL-5 ELISA standard, and standard reconstitution buffer. This matched antib 1 * 576 Tests
Catalog # ANTIA325356-696
Supplier:  ANTIBODIES.COM
New Product