ContactUsLinkComponent
This kit can be used for the detection of therapeutic antibodies, from samples obtained from experimental animals, to provide accura te pharmacokinetic data. 1 * 1 SET
Catalog # ANTIA290885-1
Supplier:  ANTIBODIES.COM
New Product
This kit can be used for the detection of therapeutic antibodies, from samples obtained from experimental animals, to provide accura te pharmacokinetic data. 1 * 1 SET
Catalog # ANTIA290885-1
Supplier:  ANTIBODIES.COM
New Product