Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
This antibody pair detects 5-Carboxylcytosine (5-caC) in denatured DNA (single strand DNA). 1 * 1 SET
Catalog # ANTIA302455-1
Supplier:
ANTIBODIES.COM
New Product
Expand image
This antibody pair detects 5-Carboxylcytosine (5-caC) in denatured DNA (single strand DNA). 1 * 1 SET
Catalog # ANTIA302455-1
Supplier:
ANTIBODIES.COM
New Product