ContactUsLinkComponent
Human Cenexin1/ODF2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determ ination of human Cenexin1/ODF2 in serum, plasma, cell culture supernatant, cell or tissue lysate, and other liquid sa 1 * 96 Tests
Catalog # ANTIA326784-96
Supplier:  Antibodies.com
New Product
Human Cenexin1/ODF2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determ ination of human Cenexin1/ODF2 in serum, plasma, cell culture supernatant, cell or tissue lysate, and other liquid sa 1 * 96 Tests
Catalog # ANTIA326784-96
Supplier:  Antibodies.com
New Product