ContactUsLinkComponent
Human Phosphoserine Aminotransferase ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qu antitative determination of human Phosphoserine Aminotransferase in serum, plasma, tissue homogenates, and other biol 1 * 96 Tests
  ANTIA302682-96
New Product
Human Phosphoserine Aminotransferase ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qu antitative determination of human Phosphoserine Aminotransferase in serum, plasma, tissue homogenates, and other biol 1 * 96 Tests
  ANTIA302682-96
New Product