ContactUsLinkComponent
Mouse IL-1 alpha ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determina tion of mouse IL-1 alpha in serum, plasma, and other biological fluids. This ELISA kit only requires 25&#0181 l of sa 1 * 96 Tests
Catalog # ANTIA323225-96
Supplier:  ANTIBODIES.COM
New Product
Mouse IL-1 alpha ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determina tion of mouse IL-1 alpha in serum, plasma, and other biological fluids. This ELISA kit only requires 25&#0181 l of sa 1 * 96 Tests
Catalog # ANTIA323225-96
Supplier:  ANTIBODIES.COM
New Product